Anti-RAP1B Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323458EA 525 EUR
ORIGTA323458
Anti-RAP1B Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:500-1000

Type: Primary
Antigen: RAP1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR