Anti-RPS6 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323449EA 506 EUR
ORIGTA323449
Anti-RPS6 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:100-200

Type: Primary
Antigen: RPS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR