Anti-SPIN2B Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323478EA 525 EUR
ORIGTA323478
Anti-SPIN2B Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000-2000; Western Blot: 1:200-1000Immunochemistry: 1:25-100

Type: Primary
Antigen: SPIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR