Anti-HDAC9 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324378EA 685 EUR
ORIGTA324378
Anti-HDAC9 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunoprecipitation: 1:100; Immunofluorescence: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: HDAC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR