Anti-USF1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324374EA 685 EUR
ORIGTA324374
Anti-USF1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50

Type: Primary
Antigen: USF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR