Anti-CDKN1C Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324373EA 735 EUR
ORIGTA324373
Anti-CDKN1C Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:1000

Type: Primary
Antigen: CDKN1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR