Anti-RORA Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324372EA 735 EUR
ORIGTA324372
Anti-RORA Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: RORA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR