Anti-PPP1R3A Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324365EA 735 EUR
ORIGTA324365
Anti-PPP1R3A Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:10-50

Type: Primary
Antigen: PPP1R3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR