Anti-EGFR Mouse Monoclonal Antibody [clone: 51CT78.40.5]

Proveedor: OriGene
ORIGTA324355EA 685 EUR
ORIGTA324355
Anti-EGFR Mouse Monoclonal Antibody [clone: 51CT78.40.5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 51CT78.40.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR