Anti-PINK1 Mouse Monoclonal Antibody [clone: 38CT20.8.5]

Proveedor: OriGene
ORIGTA324354EA 685 EUR
ORIGTA324354
Anti-PINK1 Mouse Monoclonal Antibody [clone: 38CT20.8.5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PINK1
Clonality: Monoclonal
Clone: 38CT20.8.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR