Anti-KIT Mouse Monoclonal Antibody [clone: 566CT8.5.4]

Proveedor: OriGene
ORIGTA324353EA 685 EUR
ORIGTA324353
Anti-KIT Mouse Monoclonal Antibody [clone: 566CT8.5.4]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:100

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: 566CT8.5.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR