Anti-FASN Mouse Monoclonal Antibody [clone: 497CT15.2.5]

Proveedor: OriGene
ORIGTA324351EA 735 EUR
ORIGTA324351
Anti-FASN Mouse Monoclonal Antibody [clone: 497CT15.2.5]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:200; Western Blot: 1:100-1000

Type: Primary
Antigen: FASN
Clonality: Monoclonal
Clone: 497CT15.2.5
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR