Anti-MB Mouse Monoclonal Antibody [clone: 444CT19.2.1]

Proveedor: OriGene
ORIGTA324349EA 671 EUR
ORIGTA324349
Anti-MB Mouse Monoclonal Antibody [clone: 444CT19.2.1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MB
Clonality: Monoclonal
Clone: 444CT19.2.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR