Anti-TSC2 Mouse Monoclonal Antibody [clone: 249CT19.1.3]

Proveedor: OriGene
ORIGTA324345EA 685 EUR
ORIGTA324345
Anti-TSC2 Mouse Monoclonal Antibody [clone: 249CT19.1.3]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:100

Type: Primary
Antigen: TSC2
Clonality: Monoclonal
Clone: 249CT19.1.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR