Anti-ADA Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323764EA 564 EUR
ORIGTA323764
Anti-ADA Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: ELISA: 1:1000-2000; Western Blot: 1:200-1000; Immunochemistry: 1:15-50

Type: Primary
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR