Anti-HDAC5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323445EA 564 EUR
ORIGTA323445
Anti-HDAC5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: HDAC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR