Anti-CRMP1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323439EA 564 EUR
ORIGTA323439
Anti-CRMP1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000-10000; Western Blot: 1:1000-5000

Type: Primary
Antigen: CRMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR