Anti-AKT1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323435EA 466 EUR
ORIGTA323435
Anti-AKT1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:100-200

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR