Anti-CCNC Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323428EA 506 EUR
ORIGTA323428
Anti-CCNC Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000-5000; Immunochemistry: 1:10-50

Type: Primary
Antigen: CCNC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR