Anti-MASTL Mouse Monoclonal Antibody [clone: 235CT7.8.2.3]

Proveedor: OriGene
ORIGTA324343EA 685 EUR
ORIGTA324343
Anti-MASTL Mouse Monoclonal Antibody [clone: 235CT7.8.2.3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:100-500; Immunofluorescence: 1:10-50

Type: Primary
Antigen: MASTL
Clonality: Monoclonal
Clone: 235CT7.8.2.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR