Anti-ELAVL1 Mouse Monoclonal Antibody [clone: 224CT6.5.3]

Proveedor: OriGene
ORIGTA324342EA 685 EUR
ORIGTA324342
Anti-ELAVL1 Mouse Monoclonal Antibody [clone: 224CT6.5.3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:100-1000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ELAVL1
Clonality: Monoclonal
Clone: 224CT6.5.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR