Anti-ATG4D Mouse Monoclonal Antibody [clone: 222CT15.4.1]

Proveedor: OriGene
ORIGTA324341EA 720 EUR
ORIGTA324341
Anti-ATG4D Mouse Monoclonal Antibody [clone: 222CT15.4.1]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:25; Western Blot: 1:100-1000

Type: Primary
Antigen: ATG4D
Clonality: Monoclonal
Clone: 222CT15.4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR