Anti-TRAF2 Mouse Monoclonal Antibody [clone: 214CT16.3.4]

Proveedor: OriGene
ORIGTA324340EA 685 EUR
ORIGTA324340
Anti-TRAF2 Mouse Monoclonal Antibody [clone: 214CT16.3.4]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:100-1000

Type: Primary
Antigen: TRAF2
Clonality: Monoclonal
Clone: 214CT16.3.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR