Anti-ADCK3 Mouse Monoclonal Antibody [clone: 216CT7.5.1]

Proveedor: OriGene
ORIGTA324339EA 720 EUR
ORIGTA324339
Anti-ADCK3 Mouse Monoclonal Antibody [clone: 216CT7.5.1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:100-250; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ADCK3
Clonality: Monoclonal
Clone: 216CT7.5.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR