Anti-RET Mouse Monoclonal Antibody [clone: 188CT11.2.3]

Proveedor: OriGene
ORIGTA324337EA 685 EUR
ORIGTA324337
Anti-RET Mouse Monoclonal Antibody [clone: 188CT11.2.3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-16000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: RET
Clonality: Monoclonal
Clone: 188CT11.2.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR