Anti-EIF4E Mouse Monoclonal Antibody [clone: 163CT48.1.9]

Proveedor: OriGene
ORIGTA324335EA 685 EUR
ORIGTA324335
Anti-EIF4E Mouse Monoclonal Antibody [clone: 163CT48.1.9]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:100-500

Type: Primary
Antigen: EIF4E
Clonality: Monoclonal
Clone: 163CT48.1.9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR