Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: 152CT1.2.2]

Proveedor: OriGene
ORIGTA324334EA 735 EUR
ORIGTA324334
Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: 152CT1.2.2]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:500-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: ALDH1A1
Clonality: Monoclonal
Clone: 152CT1.2.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR