Anti-EZH2 Mouse Monoclonal Antibody [clone: 144CT2.1.1.5]

Proveedor: OriGene
ORIGTA324333EA 735 EUR
ORIGTA324333
Anti-EZH2 Mouse Monoclonal Antibody [clone: 144CT2.1.1.5]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: EZH2
Clonality: Monoclonal
Clone: 144CT2.1.1.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, IgK
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR