Anti-ALDH2 Mouse Monoclonal Antibody [clone: 138CT22.3.8]

Proveedor: OriGene
ORIGTA324332EA 735 EUR
ORIGTA324332
Anti-ALDH2 Mouse Monoclonal Antibody [clone: 138CT22.3.8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ALDH2
Clonality: Monoclonal
Clone: 138CT22.3.8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR