Anti-LTF Mouse Monoclonal Antibody [clone: 119CT80.1.1]

Proveedor: OriGene
ORIGTA324329EA 685 EUR
ORIGTA324329
Anti-LTF Mouse Monoclonal Antibody [clone: 119CT80.1.1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: LTF
Clonality: Monoclonal
Clone: 119CT80.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR