Anti-MET Mouse Monoclonal Antibody [clone: 6AT1785]

Proveedor: OriGene
ORIGTA324320EA 609 EUR
ORIGTA324320
Anti-MET Mouse Monoclonal Antibody [clone: 6AT1785]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:200; Immunochemistry: 1:10-50; Western Blot: 1:100-500

Type: Primary
Antigen: MET
Clonality: Monoclonal
Clone: 6AT1785
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR