Anti-MET Mouse Monoclonal Antibody [clone: 4AT44]

Proveedor: OriGene
ORIGTA324318EA 609 EUR
ORIGTA324318
Anti-MET Mouse Monoclonal Antibody [clone: 4AT44]
Anticuerpos
Recommended Dilutions: Western Blot: 1:100-500; Immunofluorescence: 1:100; Immunochemistry: 1:50-100

Type: Primary
Antigen: MET
Clonality: Monoclonal
Clone: 4AT44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR