Anti-CPT2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323742EA 525 EUR
ORIGTA323742
Anti-CPT2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:500-5000; Western Blot: 1:500-2000; Immunochemistry: 1:25-50

Type: Primary
Antigen: CPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR