Anti-FBN1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323728EA 525 EUR
ORIGTA323728
Anti-FBN1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000-5000; Immunochemistry: 1:50-200

Type: Primary
Antigen: FBN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR