Anti-SHH Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323715EA 466 EUR
ORIGTA323715
Anti-SHH Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:2000-10000; Immunochemistry: 1:50-200

Type: Primary
Antigen: SHH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR