Anti-ITGB3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323714EA 525 EUR
ORIGTA323714
Anti-ITGB3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: ITGB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR