Anti-CDCA7L Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA325193EA 720 EUR
ORIGTA325193
Anti-CDCA7L Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CDCA7L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR