Anti-SAP30BP Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA325191EA 735 EUR
ORIGTA325191
Anti-SAP30BP Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SAP30BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR