Anti-NDUFB10 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA325189EA 684 EUR
ORIGTA325189
Anti-NDUFB10 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NDUFB10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR