Anti-BCAP31 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA325181EA 720 EUR
ORIGTA325181
Anti-BCAP31 Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: BCAP31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR