Anti-H2AFX Mouse Monoclonal Antibody [clone: 980CT1.5.5]

Proveedor: OriGene
ORIGTA325173EA 685 EUR
ORIGTA325173
Anti-H2AFX Mouse Monoclonal Antibody [clone: 980CT1.5.5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: H2AFX
Clonality: Monoclonal
Clone: 980CT1.5.5
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, zebraFish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR