Anti-KLK3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323387EA 564 EUR
ORIGTA323387
Anti-KLK3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:500-2000; Immunochemistry: 1:25-50

Type: Primary
Antigen: KLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR