Anti-ANGPTL4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323377EA 564 EUR
ORIGTA323377
Anti-ANGPTL4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:2000-5000; Immunochemistry: 1:25-100

Type: Primary
Antigen: ANGPTL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR