Anti-IL18BP Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323375EA 564 EUR
ORIGTA323375
Anti-IL18BP Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:500-5000; Immunochemistry: 1:50-200

Type: Primary
Antigen: IL18BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR