Anti-ACVR2A Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324299EA 525 EUR
ORIGTA324299
Anti-ACVR2A Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:2000-5000; Western Blot: 1:500-2000

Type: Primary
Antigen: ACVR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR