Anti-NCAM1 Mouse Monoclonal Antibody [clone: 2D4]

Proveedor: OriGene
CF506223
ORIGCF506223EA 764 EUR
ORIGCF506223
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 2D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR