Anti-LYZ Mouse Monoclonal Antibody [clone: 1D8]

Proveedor: OriGene
CF506192
ORIGCF506192EA 764 EUR
ORIGCF506192
Anti-LYZ Mouse Monoclonal Antibody [clone: 1D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: LYZ
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR