Anti-PRR11 Mouse Monoclonal Antibody [clone: 1C1]

Proveedor: OriGene
CF800542
ORIGCF800542EA 786 EUR
ORIGCF800542
Anti-PRR11 Mouse Monoclonal Antibody [clone: 1C1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PRR11
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR