Anti-PRR11 Mouse Monoclonal Antibody [clone: 3C8]

Proveedor: OriGene
ORIGCF800541EA 786 EUR
ORIGCF800541
Anti-PRR11 Mouse Monoclonal Antibody [clone: 3C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PRR11
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR