Anti-SERPINB3 Mouse Monoclonal Antibody [clone: 1F10]

Proveedor: OriGene
CF506926
ORIGCF506926EA 764 EUR
ORIGCF506926
Anti-SERPINB3 Mouse Monoclonal Antibody [clone: 1F10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: SERPINB3
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR